The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005783.3 | NP_005774.2 | 3 - 4 | 499 | 85 |
XM_017003147.1 | XP_016858636.1 | 3 - 4 | 458 | 85 | |
GenBank mRNA | AB006679.1 | - | 3 - 4 | 437 | 85 |
AB451403.1 | - | 2 - 3 | 308 | 85 | |
AK222616.1 | - | 3 - 4 | 425 | 85 | |
AK301722.1 | - | 4 - 5 | 469 | 85 | |
AK313777.1 | - | 3 - 4 | 419 | 85 | |
BC005968.1 | - | 3 - 4 | 435 | 85 | |
BC022864.1 | - | 3 - 4 | 439 | 85 | |
BC024223.2 | - | 3 - 4 | 439 | 85 | |
BC070183.1 | - | 3 - 4 | 433 | 85 | |
CR456935.1 | - | 2 - 3 | 308 | 85 |