The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305.4 | NP_001296.1 | 1 - 1 | 1248 | 129 |
GenBank mRNA | AB000712.1 | - | 1 - 1 | 1091 | 129 |
AK026651.1 | - | 1 - 1 | 1113 | 129 | |
AK124076.1 | - | 2 - 2 | 1073 | 129 | |
AK126315.1 | - | 2 - 2 | 3537 | 129 | |
AK126462.1 | - | 2 - 2 | 3589 | 129 | |
BC000671.2 | - | 1 - 1 | 1105 | 129 |