Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAN2C1
Entrez Gene ID
4123
Gene Name
mannosidase alpha class 2C member 1
Gene Aliases
MAN6A8, MANA, MANA1
Chromosome Location
Chr.15: 75355792 - 75368680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256494.1 NP_001243423.1 22 - 23 2749 73
NM_001256495.1 NP_001243424.1 22 - 23 2698 73
NM_001256496.1 NP_001243425.1 20 - 21 2401 73
NM_006715.3 NP_006706.2 22 - 23 2698 73
XM_005254384.2 XP_005254441.1 22 - 23 2751 73
XM_017022186.1 XP_016877675.1 21 - 22 3046 73
XM_017022187.1 XP_016877676.1 21 - 22 3046 73
XM_017022188.1 XP_016877677.1 20 - 21 2454 73
GenBank mRNA AF044414.2 - 22 - 23 2710 73
AK225145.1 - 20 - 21 2378 73
AL136876.1 - 22 - 23 2659 73
BC050550.1 - 22 - 23 2665 73
BC080191.1 - 22 - 23 2663 73
U37248.1 - 7 - 8 768 73

View Full Product Details