Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNDC1
Entrez Gene ID
85442
Gene Name
kinase non-catalytic C-lobe domain containing 1
Gene Aliases
C10orf23, RASGEF2, bB439H18.3
Chromosome Location
Chr.10: 133160096 - 133226412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152643.6 NP_689856.6 24 - 25 4450 70
XM_017016858.1 XP_016872347.1 24 - 25 4818 70
GenBank mRNA AK131205.1 - 5 - 6 731 70
AK294170.1 - 7 - 8 1044 70
BC047924.1 - 5 - 6 704 70
BQ719533.1 - 4 - 5 586 70

View Full Product Details