The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038618.2 | NP_001033707.1 | 7 - 8 | 1071 | 59 |
NM_001083608.1 | NP_001077077.1 | 6 - 7 | 769 | 59 | |
NM_012336.3 | NP_036468.1 | 7 - 8 | 913 | 59 | |
XM_005256340.3 | XP_005256397.1 | 7 - 8 | 1079 | 59 | |
XM_006722278.3 | XP_006722341.1 | 6 - 7 | 866 | 59 | |
XM_011523541.2 | XP_011521843.1 | 7 - 8 | 1003 | 59 | |
XM_011523543.2 | XP_011521845.1 | 8 - 9 | 916 | 59 | |
XM_017024449.1 | XP_016879938.1 | 8 - 9 | 1663 | 59 | |
XM_017024450.1 | XP_016879939.1 | 8 - 9 | 1153 | 59 | |
GenBank mRNA | AF128406.1 | - | 7 - 8 | 864 | 59 |
AK000929.1 | - | 7 - 8 | 866 | 59 | |
AK056966.1 | - | 7 - 8 | 1046 | 59 | |
AK093717.1 | - | 1 - 2 | 747 | 59 | |
AK303155.1 | - | 7 - 8 | 828 | 59 | |
AL137729.1 | - | 4 - 5 | 504 | 59 | |
BC016440.1 | - | 6 - 7 | 656 | 59 |