The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252657.1 | NP_001239586.1 | 1 - 2 | 218 | 185 |
NR_027694.1 | 1 - 2 | 218 | 185 | ||
GenBank mRNA | AK058178.1 | - | 1 - 2 | 202 | 185 |
BC105097.1 | - | 1 - 2 | 72 | 185 | |
BC105099.1 | - | 1 - 2 | 72 | 185 | |
BI463522.1 | - | 1 - 2 | 279 | 185 | |
BI463551.1 | - | 1 - 2 | 221 | 185 |