Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM14B
Entrez Gene ID
81853
Gene Name
transmembrane protein 14B
Gene Aliases
-
Chromosome Location
Chr.6: 10747759 - 10759774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127711.2 NP_001121183.1 2 - 3 188 70
NM_001286488.1 NP_001273417.1 2 - 3 188 70
NM_001286489.1 NP_001273418.1 2 - 3 188 70
NM_030969.4 NP_112231.3 2 - 3 188 70
GenBank mRNA AK291883.1 - 2 - 3 142 70
AK294152.1 - 2 - 3 142 70
AK311340.1 - 2 - 3 188 70
BC001033.1 - 2 - 3 130 70
BC007080.1 - 2 - 3 142 70
BC013913.1 - 2 - 3 150 70
BC071660.1 - 2 - 3 132 70
BG432075.1 - 2 - 3 117 70
BP249229.1 - 2 - 3 142 70
BQ056988.1 - 2 - 3 141 70
DA508140.1 - 2 - 3 186 70

View Full Product Details