Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSIP2
Entrez Gene ID
401024
Gene Name
fibrous sheath interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.2: 185738895 - 185833289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173651.3 NP_775922.3 22 - 23 20593 147
GenBank mRNA AK126036.1 - 6 - 7 2910 147
AK126051.1 - 6 - 7 4097 147
AK126089.1 - 6 - 7 3675 147
BX647691.1 - 7 - 8 3687 147
BX648733.1 - 6 - 7 4866 147

View Full Product Details