The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182640.2 | NP_872578.1 | 7 - 8 | 725 | 111 |
XM_011511644.2 | XP_011509946.1 | 6 - 7 | 565 | 111 | |
GenBank mRNA | AK130078.1 | - | 7 - 8 | 666 | 111 |
BC036575.1 | - | 8 - 9 | 678 | 111 | |
BC047784.1 | - | 6 - 7 | 375 | 111 | |
BC057240.1 | - | 7 - 8 | 679 | 111 |