Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3BP1
Entrez Gene ID
23616
Gene Name
SH3 domain binding protein 1
Gene Aliases
ARHGAP43
Chromosome Location
Chr.22: 37639677 - 37656043 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY364241.1 - 14 - 15 1500 145

View Full Product Details