The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190316.1 | NP_001177245.1 | 1 - 2 | 123 | 73 |
NM_001190317.1 | NP_001177246.1 | 1 - 2 | 123 | 73 | |
XM_006713199.3 | XP_006713262.1 | 1 - 2 | 88 | 73 | |
XM_006713200.1 | XP_006713263.1 | 1 - 2 | 61 | 73 | |
XM_006713201.1 | XP_006713264.1 | 1 - 2 | 61 | 73 | |
XM_006713202.1 | XP_006713265.1 | 1 - 2 | 66 | 73 | |
XM_017006586.1 | XP_016862075.1 | 1 - 2 | 40 | 73 | |
XM_017006587.1 | XP_016862076.1 | 1 - 2 | 40 | 73 | |
XM_017006588.1 | XP_016862077.1 | 1 - 2 | 66 | 73 | |
XM_017006589.1 | XP_016862078.1 | 1 - 2 | 86 | 73 | |
XM_017006591.1 | XP_016862080.1 | 1 - 2 | 60 | 73 | |
XM_017006592.1 | XP_016862081.1 | 1 - 2 | 86 | 73 | |
XM_017006593.1 | XP_016862082.1 | 1 - 2 | 86 | 73 | |
GenBank mRNA | AK290526.1 | - | 1 - 2 | 61 | 73 |
AK291501.1 | - | 1 - 2 | 41 | 73 | |
AK304714.1 | - | 1 - 2 | 61 | 73 | |
BX537544.1 | - | 1 - 2 | 62 | 73 |