Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
Chromosome Location
Chr.3: 138494339 - 138594383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288964.1 NP_001275893.1 11 - 12 1227 84
NM_001288965.1 NP_001275894.1 10 - 11 1171 84
NM_001288966.1 NP_001275895.1 11 - 12 1245 84
NM_001288967.1 NP_001275896.1 7 - 8 775 84
NM_001320598.1 NP_001307527.1 11 - 12 1297 84
NM_001320599.1 NP_001307528.1 11 - 12 1245 84
NM_024491.3 NP_077817.2 11 - 12 1245 84
XM_005247805.3 XP_005247862.1 9 - 10 965 84
XM_011513209.2 XP_011511511.1 9 - 10 965 84
XM_017007270.1 XP_016862759.1 9 - 10 965 84
XM_017007271.1 XP_016862760.1 11 - 12 1147 84
XM_017007272.1 XP_016862761.1 11 - 12 1485 84
XM_017007273.1 XP_016862762.1 10 - 11 1090 84
XM_017007274.1 XP_016862763.1 10 - 11 1510 84
XM_017007275.1 XP_016862764.1 11 - 12 1581 84
XM_017007276.1 XP_016862765.1 7 - 8 778 84
XM_017007277.1 XP_016862766.1 7 - 8 710 84
XM_017007278.1 XP_016862767.1 12 - 13 1316 84
XM_017007279.1 XP_016862768.1 10 - 11 1079 84
XM_017007280.1 XP_016862769.1 9 - 10 920 84
XM_017007281.1 XP_016862770.1 4 - 5 1093 84
XM_017007282.1 XP_016862771.1 10 - 11 1046 84
XM_017007283.1 XP_016862772.1 10 - 11 977 84
XM_017007284.1 XP_016862773.1 4 - 5 1281 84
GenBank mRNA AF202146.1 - 11 - 12 1099 84
AF289495.1 - 11 - 12 1099 84
AK023098.1 - 9 - 10 960 84
AK225440.1 - 11 - 12 1110 84
AK294587.1 - 7 - 8 650 84
AK299003.1 - 10 - 11 1036 84
AK301913.1 - 11 - 12 1104 84
AK314314.1 - 11 - 12 1217 84
BC030598.2 - 11 - 12 1147 84

View Full Product Details