Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
Chromosome Location
Chr.3: 138494339 - 138594383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288964.1 NP_001275893.1 8 - 9 971 145
NM_001288965.1 NP_001275894.1 7 - 8 915 145
NM_001288966.1 NP_001275895.1 8 - 9 989 145
NM_001288967.1 NP_001275896.1 4 - 5 519 145
NM_001320598.1 NP_001307527.1 8 - 9 1041 145
NM_001320599.1 NP_001307528.1 8 - 9 989 145
NM_024491.3 NP_077817.2 8 - 9 989 145
XM_005247805.3 XP_005247862.1 6 - 7 709 145
XM_011513209.2 XP_011511511.1 6 - 7 709 145
XM_017007270.1 XP_016862759.1 6 - 7 709 145
XM_017007271.1 XP_016862760.1 8 - 9 891 145
XM_017007272.1 XP_016862761.1 8 - 9 1229 145
XM_017007273.1 XP_016862762.1 7 - 8 834 145
XM_017007274.1 XP_016862763.1 7 - 8 1254 145
XM_017007275.1 XP_016862764.1 8 - 9 1325 145
XM_017007276.1 XP_016862765.1 4 - 5 522 145
XM_017007277.1 XP_016862766.1 4 - 5 454 145
XM_017007278.1 XP_016862767.1 9 - 10 1060 145
GenBank mRNA AF202146.1 - 8 - 9 843 145
AF289495.1 - 8 - 9 843 145
AK023098.1 - 6 - 7 704 145
AK225440.1 - 8 - 9 854 145
AK294587.1 - 4 - 5 394 145
AK299003.1 - 7 - 8 780 145
AK301913.1 - 8 - 9 848 145
AK314314.1 - 8 - 9 961 145
BC030598.2 - 8 - 9 891 145

View Full Product Details