The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098504.1 | NP_001091974.1 | 12 - 13 | 1791 | 90 |
GenBank mRNA | AB209595.1 | - | 12 - 13 | 1747 | 90 |
AF131750.1 | - | 1 - 2 | 1087 | 90 | |
CR456432.1 | - | 12 - 13 | 1481 | 90 |