The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136041.2 | NP_001129513.1 | 2 - 3 | 252 | 77 |
NM_024827.3 | NP_079103.2 | 2 - 3 | 284 | 77 | |
XM_005265491.1 | XP_005265548.1 | 2 - 3 | 249 | 77 | |
XM_005265492.1 | XP_005265549.1 | 2 - 3 | 328 | 77 | |
XM_006713338.1 | XP_006713401.1 | 2 - 3 | 284 | 77 | |
XM_011534131.1 | XP_011532433.1 | 2 - 3 | 372 | 77 | |
XM_011534132.2 | XP_011532434.2 | 2 - 3 | 174 | 77 | |
XM_017007234.1 | XP_016862723.1 | 2 - 3 | 372 | 77 | |
GenBank mRNA | AK025426.1 | - | 2 - 3 | 175 | 77 |
AK025890.1 | - | 2 - 3 | 175 | 77 | |
AK293223.1 | - | 2 - 3 | 185 | 77 | |
AL834223.1 | - | 2 - 3 | 542 | 77 | |
BC009676.1 | - | 1 - 2 | 160 | 77 | |
BG708402.1 | - | 2 - 3 | 254 | 77 | |
DB025431.1 | - | 2 - 3 | 284 | 77 |