The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136041.2 | NP_001129513.1 | 8 - 9 | 681 | 98 |
NM_024827.3 | NP_079103.2 | 8 - 9 | 782 | 98 | |
XM_005265491.1 | XP_005265548.1 | 8 - 9 | 747 | 98 | |
XM_005265492.1 | XP_005265549.1 | 5 - 6 | 589 | 98 | |
XM_011534131.1 | XP_011532433.1 | 8 - 9 | 870 | 98 | |
XM_011534132.2 | XP_011532434.2 | 5 - 6 | 435 | 98 | |
XM_011534135.1 | XP_011532437.1 | 7 - 8 | 589 | 98 | |
GenBank mRNA | AK025426.1 | - | 8 - 9 | 673 | 98 |
AK025890.1 | - | 8 - 9 | 673 | 98 | |
AK293223.1 | - | 8 - 9 | 614 | 98 | |
AK299021.1 | - | 4 - 5 | 353 | 98 | |
AL834223.1 | - | 7 - 8 | 997 | 98 | |
BC009676.1 | - | 7 - 8 | 658 | 98 | |
CR457341.1 | - | 4 - 5 | 283 | 98 |