Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHA3
Entrez Gene ID
65977
Gene Name
pleckstrin homology domain containing A3
Gene Aliases
FAPP1
Chromosome Location
Chr.2: 178480410 - 178516324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011511689.2 XP_011509991.1 2 - 3 616 110

View Full Product Details