Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHA3
Entrez Gene ID
65977
Gene Name
pleckstrin homology domain containing A3
Gene Aliases
FAPP1
Chromosome Location
Chr.2: 178480410 - 178516324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019091.3 NP_061964.3 3 - 4 712 140
XM_011511689.2 XP_011509991.1 4 - 5 821 140
GenBank mRNA AF286162.1 - 3 - 4 551 140
AK000074.1 - 1 - 2 115 140
AK074209.1 - 3 - 4 687 140
AK314459.1 - 3 - 4 716 140
BC044567.1 - 3 - 4 406 140

View Full Product Details