The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014648.3 | NP_055463.1 | 23 - 24 | 3010 | 65 |
XM_005247914.3 | XP_005247971.1 | 23 - 24 | 2809 | 65 | |
XM_005247915.3 | XP_005247972.1 | 23 - 24 | 2779 | 65 | |
XM_005247916.3 | XP_005247973.1 | 24 - 25 | 2951 | 65 | |
XM_005247917.3 | XP_005247974.1 | 23 - 24 | 2740 | 65 | |
XM_005247918.3 | XP_005247975.1 | 23 - 24 | 3227 | 65 | |
XM_005247919.3 | XP_005247976.1 | 22 - 23 | 2361 | 65 | |
XM_017007540.1 | XP_016863029.1 | 23 - 24 | 2856 | 65 | |
GenBank mRNA | AB014575.1 | - | 23 - 24 | 2717 | 65 |
AF279370.1 | - | 8 - 9 | 669 | 65 | |
AK023138.1 | - | 23 - 24 | 2778 | 65 | |
AK294967.1 | - | 12 - 13 | 1073 | 65 | |
AK297451.1 | - | 9 - 10 | 775 | 65 | |
AK303942.1 | - | 10 - 11 | 1213 | 65 | |
AY227652.1 | - | 10 - 11 | 921 | 65 | |
AY227654.1 | - | 22 - 23 | 3586 | 65 | |
BC063882.1 | - | 23 - 24 | 2763 | 65 |