The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014648.3 | NP_055463.1 | 31 - 32 | 3936 | 69 |
XM_005247914.3 | XP_005247971.1 | 31 - 32 | 3735 | 69 | |
XM_005247915.3 | XP_005247972.1 | 31 - 32 | 3705 | 69 | |
XM_005247916.3 | XP_005247973.1 | 32 - 33 | 3877 | 69 | |
XM_005247917.3 | XP_005247974.1 | 31 - 32 | 3666 | 69 | |
XM_005247918.3 | XP_005247975.1 | 31 - 32 | 4153 | 69 | |
XM_005247919.3 | XP_005247976.1 | 30 - 31 | 3287 | 69 | |
XM_017007540.1 | XP_016863029.1 | 31 - 32 | 3782 | 69 | |
GenBank mRNA | AB014575.1 | - | 31 - 32 | 3643 | 69 |
AF279370.1 | - | 16 - 17 | 1594 | 69 | |
AK023138.1 | - | 31 - 32 | 3704 | 69 | |
AK294967.1 | - | 20 - 21 | 1999 | 69 | |
AY227652.1 | - | 18 - 19 | 1847 | 69 | |
AY227654.1 | - | 29 - 30 | 4372 | 69 | |
AY495710.1 | - | 6 - 7 | 717 | 69 | |
BC063882.1 | - | 31 - 32 | 3689 | 69 |