The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017906.2 | NP_060376.2 | 4 - 5 | 475 | 109 |
XM_005249204.2 | XP_005249261.1 | 5 - 6 | 868 | 109 | |
XM_011514720.1 | XP_011513022.1 | 5 - 6 | 528 | 109 | |
XM_011514721.1 | XP_011513023.1 | 5 - 6 | 525 | 109 | |
GenBank mRNA | AF283303.1 | - | 4 - 5 | 518 | 109 |
AK000631.1 | - | 4 - 5 | 523 | 109 | |
AK225516.1 | - | 4 - 5 | 523 | 109 | |
BC010907.1 | - | 4 - 5 | 479 | 109 | |
KU178675.1 | - | 4 - 5 | 444 | 109 | |
KU178676.1 | - | 3 - 4 | 281 | 109 |