Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HP
Entrez Gene ID
3240
Gene Name
haptoglobin
Gene Aliases
BP, HP2ALPHA2, HPA1S
Chromosome Location
Chr.16: 72054592 - 72061056 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005143.4 NP_005134.1 4 - 5 331 91
GenBank mRNA AK314700.1 - 4 - 5 300 91
BC121125.1 - 4 - 5 294 91
CB134891.1 - 4 - 5 314 91
L29394.1 - 4 - 5 296 91

View Full Product Details