The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173156.2 | NP_775179.1 | 1 - 2 | 276 | 67 |
NM_201568.2 | NP_963862.1 | 1 - 2 | 276 | 67 | |
NM_201569.2 | NP_963863.2 | 1 - 2 | 276 | 67 | |
XM_005245652.3 | XP_005245709.1 | 1 - 2 | 145 | 67 | |
XM_006711678.3 | XP_006711741.1 | 1 - 2 | 145 | 67 | |
XM_017002969.1 | XP_016858458.1 | 1 - 2 | 276 | 67 | |
XM_017002970.1 | XP_016858459.1 | 1 - 2 | 276 | 67 | |
XM_017002971.1 | XP_016858460.1 | 1 - 2 | 276 | 67 | |
GenBank mRNA | AB085674.1 | - | 1 - 2 | 89 | 67 |
AK056035.1 | - | 1 - 2 | 145 | 67 | |
AK074297.1 | - | 1 - 2 | 133 | 67 | |
AK307394.1 | - | 1 - 2 | 137 | 67 | |
BC036381.1 | - | 1 - 2 | 144 | 67 | |
BM152501.1 | - | 1 - 2 | 149 | 67 | |
D87437.2 | - | 1 - 2 | 120 | 67 | |
DB223038.1 | - | 1 - 2 | 276 | 67 | |
DC380923.1 | - | 1 - 2 | 258 | 67 |