Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMG7
Entrez Gene ID
9887
Gene Name
SMG7, nonsense mediated mRNA decay factor
Gene Aliases
C1orf16, EST1C, SGA56M
Chromosome Location
Chr.1: 183471962 - 183554193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174061.1 NP_001167532.1 2 - 3 398 141
NM_173156.2 NP_775179.1 3 - 4 430 141
NM_201568.2 NP_963862.1 3 - 4 430 141
NM_201569.2 NP_963863.2 3 - 4 430 141
XM_005245648.3 XP_005245705.1 5 - 6 1115 141
XM_005245649.3 XP_005245706.1 4 - 5 978 141
XM_005245652.3 XP_005245709.1 3 - 4 299 141
XM_005245653.4 XP_005245710.1 4 - 5 401 141
XM_006711676.3 XP_006711739.1 4 - 5 978 141
XM_006711678.3 XP_006711741.1 3 - 4 299 141
XM_011510205.2 XP_011508507.1 6 - 7 1293 141
XM_011510206.2 XP_011508508.1 6 - 7 1293 141
XM_011510207.2 XP_011508509.1 4 - 5 1017 141
XM_011510208.2 XP_011508510.1 6 - 7 1293 141
XM_011510209.2 XP_011508511.1 6 - 7 1293 141
XM_011510210.2 XP_011508512.1 2 - 3 269 141
XM_017002966.1 XP_016858455.1 5 - 6 570 141
XM_017002967.1 XP_016858456.1 5 - 6 578 141
XM_017002968.1 XP_016858457.1 2 - 3 273 141
GenBank mRNA AK056035.1 - 3 - 4 299 141
AK091477.1 - 2 - 3 267 141
AK297207.1 - 4 - 5 462 141
AK299178.1 - 2 - 3 269 141
AK307394.1 - 3 - 4 291 141
BC036381.1 - 3 - 4 298 141
D87437.2 - 3 - 4 274 141
DC380923.1 - 3 - 4 412 141

View Full Product Details