Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAKD
Entrez Gene ID
79877
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
-
Chromosome Location
Chr.17: 45023338 - 45061129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128631.2 NP_001122103.1 4 - 5 628 72
NM_001288654.2 NP_001275583.1 4 - 5 764 72
NM_001288655.1 NP_001275584.1 4 - 5 676 72
NM_001321326.1 NP_001308255.1 4 - 5 574 72
NM_024819.6 NP_079095.3 4 - 5 761 72
XM_011525262.2 XP_011523564.1 4 - 5 793 72
XM_017025105.1 XP_016880594.1 5 - 6 866 72
XM_017025106.1 XP_016880595.1 5 - 6 808 72
GenBank mRNA AK290755.1 - 4 - 5 577 72
BC006472.1 - 4 - 5 650 72
BC006546.2 - 4 - 5 650 72
BC018132.2 - 3 - 4 522 72
BC033299.1 - 3 - 4 661 72
BM910634.1 - 4 - 5 675 72
DR005747.1 - 4 - 5 676 72

View Full Product Details