The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152643.6 | NP_689856.6 | 14 - 15 | 2753 | 86 |
XM_017016858.1 | XP_016872347.1 | 14 - 15 | 3121 | 86 | |
XM_017016859.1 | XP_016872348.1 | 14 - 15 | 3124 | 86 | |
GenBank mRNA | AB000781.1 | - | 13 - 14 | 2649 | 86 |
AB051555.1 | - | 13 - 14 | 2847 | 86 | |
AK074179.1 | - | 4 - 5 | 1019 | 86 | |
AK090457.1 | - | 13 - 14 | 3660 | 86 |