Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNDC1
Entrez Gene ID
85442
Gene Name
kinase non-catalytic C-lobe domain containing 1
Gene Aliases
C10orf23, RASGEF2, bB439H18.3
Chromosome Location
Chr.10: 133160096 - 133226412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152643.6 NP_689856.6 15 - 16 2903 90
XM_017016858.1 XP_016872347.1 15 - 16 3271 90
XM_017016859.1 XP_016872348.1 15 - 16 3274 90
GenBank mRNA AB051555.1 - 14 - 15 2997 90
AK074179.1 - 5 - 6 1169 90
AK090457.1 - 14 - 15 3810 90

View Full Product Details