Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMG1
Entrez Gene ID
23049
Gene Name
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Gene Aliases
61E3.4, ATX, LIP
Chromosome Location
Chr.16: 18804853 - 18926417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015092.4 NP_055907.3 34 - 35 5644 83
XM_005255182.3 XP_005255239.1 34 - 35 5657 83
XM_005255183.3 XP_005255240.1 35 - 36 5715 83
XM_005255184.3 XP_005255241.1 33 - 34 5501 83
XM_011545769.2 XP_011544071.1 34 - 35 5657 83
XM_011545770.1 XP_011544072.1 22 - 23 3454 83
XM_017023066.1 XP_016878555.1 34 - 35 5467 83
XM_017023067.1 XP_016878556.1 22 - 23 3517 83
GenBank mRNA AB007881.2 - 2 - 3 262 83
AB061371.1 - 35 - 36 5596 83
AF186377.1 - 9 - 10 1625 83
AF395444.1 - 32 - 33 4951 83
AK074359.1 - 1 - 2 123 83
AY014957.1 - 22 - 23 3523 83

View Full Product Details