The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015092.4 | NP_055907.3 | 41 - 42 | 7060 | 62 |
XM_005255182.3 | XP_005255239.1 | 41 - 42 | 7073 | 62 | |
XM_005255183.3 | XP_005255240.1 | 42 - 43 | 7131 | 62 | |
XM_005255184.3 | XP_005255241.1 | 40 - 41 | 6917 | 62 | |
XM_011545769.2 | XP_011544071.1 | 41 - 42 | 7073 | 62 | |
XM_011545770.1 | XP_011544072.1 | 29 - 30 | 4870 | 62 | |
XM_017023066.1 | XP_016878555.1 | 41 - 42 | 6883 | 62 | |
XM_017023067.1 | XP_016878556.1 | 29 - 30 | 4933 | 62 | |
GenBank mRNA | AB007881.2 | - | 9 - 10 | 1678 | 62 |
AB061371.1 | - | 42 - 43 | 7012 | 62 | |
AF186377.1 | - | 16 - 17 | 3041 | 62 | |
AF395444.1 | - | 39 - 40 | 6367 | 62 | |
AK074359.1 | - | 8 - 9 | 1539 | 62 | |
AL833298.1 | - | 2 - 3 | 399 | 62 | |
AY014957.1 | - | 29 - 30 | 4939 | 62 |