The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015092.4 | NP_055907.3 | 52 - 53 | 9306 | 87 |
XM_005255182.3 | XP_005255239.1 | 51 - 52 | 9403 | 87 | |
XM_005255183.3 | XP_005255240.1 | 52 - 53 | 9461 | 87 | |
XM_005255184.3 | XP_005255241.1 | 50 - 51 | 9247 | 87 | |
XM_011545769.2 | XP_011544071.1 | 51 - 52 | 9403 | 87 | |
XM_011545770.1 | XP_011544072.1 | 39 - 40 | 7200 | 87 | |
XM_017023066.1 | XP_016878555.1 | 51 - 52 | 9213 | 87 | |
XM_017023067.1 | XP_016878556.1 | 39 - 40 | 7263 | 87 | |
GenBank mRNA | AB007881.2 | - | 20 - 21 | 3924 | 87 |
AB061371.1 | - | 53 - 54 | 9258 | 87 | |
AF186377.1 | - | 27 - 28 | 5287 | 87 | |
AF395444.1 | - | 50 - 51 | 8613 | 87 | |
AL833298.1 | - | 13 - 14 | 2644 | 87 | |
AY014957.1 | - | 40 - 41 | 7185 | 87 | |
U32581.2 | - | 2 - 3 | 316 | 87 |