Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXB6
Entrez Gene ID
3216
Gene Name
homeobox B6
Gene Aliases
HOX2, HOX2B, HU-2, Hox-2.2
Chromosome Location
Chr.17: 48595736 - 48604992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018952.4 NP_061825.2 2 - 3 245 98
XM_005257283.3 XP_005257340.1 1 - 2 487 98
GenBank mRNA BC014651.1 - 2 - 3 245 98

View Full Product Details