Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP95
Entrez Gene ID
90799
Gene Name
centrosomal protein 95
Gene Aliases
CCDC45
Chromosome Location
Chr.17: 64506736 - 64537951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316990.1 NP_001303919.1 1 - 2 380 117
NM_138363.2 NP_612372.1 1 - 2 380 117
NR_133644.1 1 - 2 380 117
XM_005257779.3 XP_005257836.1 1 - 2 356 117
XM_005257780.2 XP_005257837.1 1 - 2 105 117
XM_006722166.3 XP_006722229.1 1 - 2 122 117
XM_011525458.2 XP_011523760.1 1 - 2 356 117
XM_017025287.1 XP_016880776.1 1 - 2 124 117
XM_017025288.1 XP_016880777.1 1 - 2 122 117
GenBank mRNA AK057326.1 - 1 - 2 354 117
AK290587.1 - 1 - 2 120 117
AK297409.1 - 1 - 2 81 117
AK316257.1 - 1 - 2 124 117
BC009518.1 - 1 - 2 76 117
BP357015.1 - 1 - 2 380 117

View Full Product Details