Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP95
Entrez Gene ID
90799
Gene Name
centrosomal protein 95
Gene Aliases
CCDC45
Chromosome Location
Chr.17: 64506736 - 64537951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316990.1 NP_001303919.1 8 - 9 1282 65
NM_138363.2 NP_612372.1 9 - 10 1388 65
NR_133644.1 9 - 10 1407 65
XM_005257779.3 XP_005257836.1 9 - 10 1364 65
XM_005257780.2 XP_005257837.1 8 - 9 1007 65
XM_006722166.3 XP_006722229.1 9 - 10 1146 65
XM_011525458.2 XP_011523760.1 9 - 10 1364 65
XM_017025287.1 XP_016880776.1 9 - 10 2367 65
XM_017025288.1 XP_016880777.1 8 - 9 1040 65
GenBank mRNA AK057326.1 - 9 - 10 1362 65
AK297409.1 - 8 - 9 983 65
AL832822.1 - 9 - 10 1058 65
BC009518.1 - 9 - 10 1084 65

View Full Product Details