Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TNFRSF25
- Entrez Gene ID
-
8718
- Gene Name
- TNF receptor superfamily member 25
- Gene Aliases
-
APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LR
- Chromosome Location
-
Chr.1: 6461151 - 6466195 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U94507.1 |
- |
3 - 4 |
324 |
118 |
View Full Product Details