The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039141.2 | NP_001034230.1 | 10 - 11 | 5454 | 52 |
GenBank mRNA | DQ228003.1 | - | 8 - 9 | 5117 | 52 |
DQ228005.1 | - | 9 - 10 | 5319 | 52 |