The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006724174.3 | XP_006724237.1 | 3 - 4 | 413 | 80 |
XM_006724175.3 | XP_006724238.1 | 3 - 4 | 415 | 80 | |
XM_006724176.3 | XP_006724239.1 | 3 - 4 | 415 | 80 | |
XM_006724178.3 | XP_006724241.1 | 3 - 4 | 415 | 80 | |
XM_011530025.2 | XP_011528327.1 | 3 - 4 | 419 | 80 |