The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003940.2 | NP_003931.2 | 11 - 12 | 1463 | 100 |
XM_011513269.1 | XP_011511571.1 | 11 - 12 | 1410 | 100 | |
XM_017007425.1 | XP_016862914.1 | 11 - 12 | 1307 | 100 | |
XM_017007426.1 | XP_016862915.1 | 10 - 11 | 1274 | 100 | |
XM_017007427.1 | XP_016862916.1 | 8 - 9 | 994 | 100 | |
GenBank mRNA | AK290338.1 | - | 11 - 12 | 1410 | 100 |
AK302404.1 | - | 11 - 12 | 1272 | 100 | |
BC016146.1 | - | 11 - 12 | 1416 | 100 | |
HM138083.1 | - | 11 - 12 | 1382 | 100 | |
U75362.1 | - | 11 - 12 | 1476 | 100 |