The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003940.2 | NP_003931.2 | 1 - 2 | 247 | 119 |
XM_011513269.1 | XP_011511571.1 | 1 - 2 | 194 | 119 | |
GenBank mRNA | AK290338.1 | - | 1 - 2 | 194 | 119 |
BC016146.1 | - | 1 - 2 | 200 | 119 | |
HM138083.1 | - | 1 - 2 | 166 | 119 | |
U75362.1 | - | 1 - 2 | 260 | 119 |