The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003940.2 | NP_003931.2 | 4 - 5 | 557 | 147 |
XM_011513269.1 | XP_011511571.1 | 4 - 5 | 504 | 147 | |
XM_017007425.1 | XP_016862914.1 | 4 - 5 | 401 | 147 | |
XM_017007426.1 | XP_016862915.1 | 3 - 4 | 368 | 147 | |
GenBank mRNA | AK290338.1 | - | 4 - 5 | 504 | 147 |
AK302404.1 | - | 4 - 5 | 366 | 147 | |
BC016146.1 | - | 4 - 5 | 510 | 147 | |
HM138083.1 | - | 4 - 5 | 476 | 147 | |
U75362.1 | - | 4 - 5 | 570 | 147 |