The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015046.5 | NP_055861.3 | 19 - 20 | 6728 | 79 |
XM_005272171.1 | XP_005272228.1 | 19 - 20 | 6729 | 79 | |
XM_005272172.2 | XP_005272229.1 | 20 - 21 | 6827 | 79 | |
XM_005272173.2 | XP_005272230.1 | 19 - 20 | 6865 | 79 | |
XM_011518404.2 | XP_011516706.1 | 20 - 21 | 6869 | 79 | |
XM_011518405.2 | XP_011516707.1 | 19 - 20 | 6887 | 79 | |
XM_011518406.2 | XP_011516708.1 | 19 - 20 | 6729 | 79 | |
XM_011518407.1 | XP_011516709.1 | 19 - 20 | 6729 | 79 | |
XM_017014495.1 | XP_016869984.1 | 19 - 20 | 6800 | 79 | |
XM_017014496.1 | XP_016869985.1 | 10 - 11 | 1396 | 79 | |
XM_017014497.1 | XP_016869986.1 | 10 - 11 | 1765 | 79 | |
GenBank mRNA | AB014525.2 | - | 17 - 18 | 6505 | 79 |
AK001456.1 | - | 9 - 10 | 1264 | 79 | |
AK025601.1 | - | 4 - 5 | 359 | 79 | |
AK226007.1 | - | 10 - 11 | 1314 | 79 | |
AY362728.1 | - | 17 - 18 | 6552 | 79 | |
BC032600.2 | - | 10 - 11 | 1305 | 79 | |
BC032622.1 | - | 10 - 11 | 1606 | 79 | |
BC078166.1 | - | 9 - 10 | 1265 | 79 | |
BC106017.1 | - | 8 - 9 | 1116 | 79 | |
BC137350.1 | - | 18 - 19 | 6662 | 79 | |
BX537849.1 | - | 19 - 20 | 6713 | 79 | |
BX538166.1 | - | 19 - 20 | 6731 | 79 | |
CR749249.1 | - | 10 - 11 | 5209 | 79 | |
KU178559.1 | - | 7 - 8 | 998 | 79 | |
KU178560.1 | - | 7 - 8 | 998 | 79 |