The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037324.2 | NP_001032401.1 | 2 - 3 | 322 | 84 |
NM_001100120.1 | NP_001093590.1 | 3 - 4 | 547 | 84 | |
NM_001100121.1 | NP_001093591.1 | 3 - 4 | 409 | 84 | |
NM_014693.3 | NP_055508.3 | 3 - 4 | 658 | 84 | |
GenBank mRNA | AB011176.1 | - | 3 - 4 | 404 | 84 |
AF192531.1 | - | 3 - 4 | 314 | 84 | |
AF428263.1 | - | 3 - 4 | 586 | 84 | |
AF428264.1 | - | 2 - 3 | 227 | 84 | |
AF521189.1 | - | 3 - 4 | 523 | 84 | |
AK293713.1 | - | 3 - 4 | 428 | 84 | |
AK293891.1 | - | 2 - 3 | 252 | 84 | |
AK296541.1 | - | 2 - 3 | 305 | 84 | |
AK309236.1 | - | 1 - 2 | 102 | 84 | |
AY359003.1 | - | 2 - 3 | 278 | 84 | |
BC142950.1 | - | 2 - 3 | 322 | 84 |