The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037324.2 | NP_001032401.1 | 8 - 9 | 1199 | 68 |
NM_001100120.1 | NP_001093590.1 | 9 - 10 | 1424 | 68 | |
NM_001100121.1 | NP_001093591.1 | 9 - 10 | 1286 | 68 | |
NM_014693.3 | NP_055508.3 | 9 - 10 | 1535 | 68 | |
GenBank mRNA | AB011176.1 | - | 9 - 10 | 1281 | 68 |
AF192531.1 | - | 9 - 10 | 1191 | 68 | |
AF428263.1 | - | 9 - 10 | 1463 | 68 | |
AF428264.1 | - | 8 - 9 | 1104 | 68 | |
AF521189.1 | - | 9 - 10 | 1400 | 68 | |
AK293891.1 | - | 8 - 9 | 1129 | 68 | |
AK295270.1 | - | 2 - 3 | 336 | 68 | |
AK296541.1 | - | 8 - 9 | 1182 | 68 | |
AK309236.1 | - | 6 - 7 | 913 | 68 | |
AY359003.1 | - | 8 - 9 | 1155 | 68 | |
BC142950.1 | - | 8 - 9 | 1199 | 68 |