The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014693.3 | NP_055508.3 | 2 - 3 | 513 | 67 |
GenBank mRNA | AF428263.1 | - | 2 - 3 | 441 | 67 |