Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECE2
Entrez Gene ID
9718
Gene Name
endothelin converting enzyme 2
Gene Aliases
-
Chromosome Location
Chr.3: 184249657 - 184293031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037324.2 NP_001032401.1 1 - 2 188 67
GenBank mRNA AF428264.1 - 1 - 2 93 67
AK296541.1 - 1 - 2 171 67
AY359003.1 - 1 - 2 144 67
BC142950.1 - 1 - 2 188 67

View Full Product Details