Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP70
Entrez Gene ID
80321
Gene Name
centrosomal protein 70
Gene Aliases
BITE
Chromosome Location
Chr.3: 138494339 - 138594383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288964.1 NP_001275893.1 5 - 6 564 96
NM_001288965.1 NP_001275894.1 4 - 5 508 96
NM_001288966.1 NP_001275895.1 5 - 6 582 96
NM_001320598.1 NP_001307527.1 5 - 6 634 96
NM_001320599.1 NP_001307528.1 5 - 6 582 96
NM_001320600.1 NP_001307529.1 4 - 5 448 96
NM_024491.3 NP_077817.2 5 - 6 582 96
XM_005247805.3 XP_005247862.1 3 - 4 302 96
XM_011513209.2 XP_011511511.1 3 - 4 302 96
XM_017007270.1 XP_016862759.1 3 - 4 302 96
XM_017007271.1 XP_016862760.1 5 - 6 484 96
XM_017007272.1 XP_016862761.1 5 - 6 822 96
XM_017007273.1 XP_016862762.1 4 - 5 427 96
XM_017007274.1 XP_016862763.1 4 - 5 847 96
XM_017007275.1 XP_016862764.1 5 - 6 918 96
XM_017007278.1 XP_016862767.1 5 - 6 554 96
XM_017007279.1 XP_016862768.1 4 - 5 430 96
XM_017007280.1 XP_016862769.1 4 - 5 427 96
XM_017007282.1 XP_016862771.1 5 - 6 553 96
XM_017007283.1 XP_016862772.1 5 - 6 484 96
GenBank mRNA AF202146.1 - 5 - 6 436 96
AF289495.1 - 5 - 6 436 96
AK023098.1 - 3 - 4 297 96
AK225440.1 - 5 - 6 447 96
AK295866.1 - 5 - 6 453 96
AK299003.1 - 4 - 5 373 96
AK301913.1 - 5 - 6 441 96
AK314314.1 - 5 - 6 554 96
BC016050.2 - 4 - 5 449 96
BC030598.2 - 5 - 6 484 96
BM908227.1 - - 444 96
BX331994.2 - 5 - 6 591 96

View Full Product Details