Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDEM3
Entrez Gene ID
80267
Gene Name
ER degradation enhancing alpha-mannosidase like protein 3
Gene Aliases
C1orf22
Chromosome Location
Chr.1: 184690491 - 184754907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319960.1 NP_001306889.1 7 - 8 1014 152
NM_025191.3 NP_079467.3 7 - 8 1014 152
NR_135118.1 7 - 8 1014 152
XM_005245499.1 XP_005245556.1 7 - 8 995 152
XM_011510012.1 XP_011508314.1 6 - 7 934 152
XM_011510013.2 XP_011508315.1 7 - 8 995 152
XM_011510014.2 XP_011508316.1 7 - 8 995 152
XM_017002397.1 XP_016857886.1 6 - 7 934 152
XM_017002398.1 XP_016857887.1 6 - 7 934 152
GenBank mRNA AF288393.1 - 7 - 8 677 152
AK225171.1 - 7 - 8 957 152
AK315118.1 - 7 - 8 958 152
BC105586.1 - 6 - 7 910 152
BC144149.1 - 7 - 8 674 152
BC144150.1 - 7 - 8 674 152

View Full Product Details