The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020376.3 | NP_065109.1 | 6 - 7 | 958 | 109 |
XM_006718265.3 | XP_006718328.1 | 6 - 7 | 964 | 109 | |
XM_006718266.3 | XP_006718329.1 | 5 - 6 | 904 | 109 | |
XM_017018028.1 | XP_016873517.1 | 5 - 6 | 904 | 109 | |
GenBank mRNA | AF055000.1 | - | 4 - 5 | 476 | 109 |
AJ278475.1 | - | 6 - 7 | 827 | 109 | |
AJ278476.1 | - | 5 - 6 | 847 | 109 | |
AL833707.1 | - | 1 - 2 | 43 | 109 | |
AY894804.1 | - | 5 - 6 | 755 | 109 | |
BC011958.1 | - | 3 - 4 | 346 | 109 | |
BC017280.2 | - | 6 - 7 | 957 | 109 | |
HQ651812.1 | - | 6 - 7 | 958 | 109 | |
JF279441.1 | - | 6 - 7 | 958 | 109 | |
JF279442.1 | - | 6 - 7 | 958 | 109 | |
KF601369.1 | - | 6 - 7 | 958 | 109 | |
KF833264.1 | - | 5 - 6 | 755 | 109 | |
KF833265.1 | - | 5 - 6 | 755 | 109 |