The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004645.2 | NP_004636.1 | 3 - 4 | 1473 | 86 |
GenBank mRNA | AK313616.1 | - | 3 - 4 | 1455 | 86 |
BC010385.1 | - | 3 - 4 | 1470 | 86 | |
M58411.1 | - | 2 - 3 | 930 | 86 | |
U06632.1 | - | 3 - 4 | 1464 | 86 |