The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317724.1 | NP_001304653.1 | 4 - 5 | 443 | 152 |
NM_016167.4 | NP_057251.2 | 4 - 5 | 443 | 152 | |
GenBank mRNA | AF172066.3 | - | 4 - 5 | 443 | 152 |
AK075301.1 | - | 4 - 5 | 426 | 152 | |
AL050299.1 | - | 3 - 4 | 143 | 152 | |
BC023517.2 | - | 4 - 5 | 418 | 152 | |
BC062683.1 | - | 4 - 5 | 439 | 152 | |
BU071935.1 | - | 4 - 5 | 196 | 152 |