The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317724.1 | NP_001304653.1 | 6 - 7 | 650 | 107 |
NM_016167.4 | NP_057251.2 | 6 - 7 | 650 | 107 | |
GenBank mRNA | AF172066.3 | - | 6 - 7 | 650 | 107 |
AK075301.1 | - | 6 - 7 | 633 | 107 | |
BC023517.2 | - | 6 - 7 | 625 | 107 | |
BC062683.1 | - | 6 - 7 | 646 | 107 | |
BU071935.1 | - | 6 - 7 | 403 | 107 |